Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482288.4 | XP_003482336.1 | 203 | 64 | ||
| GenBank mRNA | AK352499.1 | - | 2 - 3 | 188 | 64 | |
| AK391975.1 | - | 2 - 3 | 184 | 64 | ||
| AY610003.1 | - | 2 - 3 | 182 | 64 | ||
More Information
Set Membership: |
|

ISCA2
Genomic Map