Assay Details
| Gene Symbol: DCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123832.5 | XP_003123880.1 | 1030 | 71 | ||
| XM_013995033.2 | XP_013850487.1 | 951 | 71 | |||
| GenBank mRNA | AK231565.1 | - | 5 - 6 | 575 | 71 | |
| AK347285.1 | - | 7 - 8 | 898 | 71 | ||
| AK400628.1 | - | 7 - 8 | 953 | 71 | ||
More Information
Set Membership: |
|

DCP2
Genomic Map