Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653760.3 | XP_005653817.1 | 559 | 98 | ||
| XM_005653761.3 | XP_005653818.1 | 560 | 98 | |||
| XM_013978068.2 | XP_013833522.1 | 560 | 98 | |||
| XM_021064300.1 | XP_020919959.1 | 560 | 98 | |||
| GenBank mRNA | AK231881.1 | - | 3 - 4 | 549 | 98 | |
More Information
Set Membership: |
|

RRP15
Genomic Map