Assay Details
| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996824.2 | XP_013852278.1 | 661 | 104 | ||
| XM_013996825.2 | XP_013852279.1 | 527 | 104 | |||
| GenBank mRNA | AK231912.1 | - | 5 - 6 | 559 | 104 | |
More Information
Set Membership: |
|

CFAP126
Genomic Map