Assay Details
| Gene Symbol: TMEM167B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926138.5 | XP_001926173.1 | 102 | |||
| GenBank mRNA | AK389608.1 | - | 1 | 1018 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM167B| Gene Symbol: TMEM167B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926138.5 | XP_001926173.1 | 102 | |||
| GenBank mRNA | AK389608.1 | - | 1 | 1018 | 102 | |
Set Membership: |
|