Assay Details
| Gene Symbol: LHX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021068367.1 | XP_020924026.1 | 794 | 106 | ||
| XM_021068376.1 | XP_020924035.1 | 796 | 106 | |||
| XM_021068384.1 | XP_020924043.1 | 553 | 106 | |||
| XM_021068392.1 | XP_020924051.1 | 260 | 106 | |||
| GenBank mRNA | AK390019.1 | - | 2 - 3 | 332 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021068392.1 transcripts.
Set Membership: |
|

LHX6
Genomic Map