Assay Details
| Gene Symbol: CELF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128507.5 | XP_003128555.2 | 827 | 62 | ||
| GenBank mRNA | AK390459.1 | - | 4 - 5 | 633 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CELF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128507.5 | XP_003128555.2 | 827 | 62 | ||
| GenBank mRNA | AK390459.1 | - | 4 - 5 | 633 | 62 | |
Set Membership: |
|