Assay Details
| Gene Symbol: RNF186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095470.1 | XP_020951129.1 | 1393 | 71 | ||
| GenBank mRNA | AK390872.1 | - | 1 | 1020 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RNF186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095470.1 | XP_020951129.1 | 1393 | 71 | ||
| GenBank mRNA | AK390872.1 | - | 1 | 1020 | 71 | |
Set Membership: |
|