Assay Details
| Gene Symbol: STUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124708.5 | XP_003124756.2 | 790 | 70 | ||
| GenBank mRNA | AK391602.1 | - | 5 - 6 | 766 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: STUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124708.5 | XP_003124756.2 | 790 | 70 | ||
| GenBank mRNA | AK391602.1 | - | 5 - 6 | 766 | 70 | |
Set Membership: |
|