Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130090.4 | XP_003130138.2 | 1573 | 58 | ||
| XM_013979603.1 | XP_013835057.1 | 1245 | 58 | |||
| GenBank mRNA | AK391610.1 | - | 6 - 7 | 1525 | 58 | |
More Information
Set Membership: |
|

B3GAT1
Genomic Map