Assay Details
| Gene Symbol: SEC61G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005667909.3 | XP_005667966.1 | 567 | 81 | ||
| XM_021063933.1 | XP_020919592.1 | 564 | 81 | |||
| GenBank mRNA | AK391814.1 | - | 2 - 3 | 417 | 81 | |
More Information
Set Membership: |
|

SEC61G
Genomic Map