Assay Details
| Gene Symbol: GABRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356906.4 | XP_003356954.1 | 3505 | 106 | ||
| XM_021101169.1 | XP_020956828.1 | 3314 | 106 | |||
| GenBank mRNA | AK392013.1 | - | 1 | 723 | 106 | |
More Information
Set Membership: |
|

GABRA4
Genomic Map