Assay Details
| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091430.1 | XP_020947089.1 | 819 | 64 | ||
| XM_021091431.1 | XP_020947090.1 | 819 | 64 | |||
| XM_021091432.1 | XP_020947091.1 | 876 | 64 | |||
| GenBank mRNA | AK392686.1 | - | 6 - 7 | 767 | 64 | |
More Information
Set Membership: |
|

RRP7A
Genomic Map