Assay Details
| Gene Symbol: SPRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087442.1 | XP_020943101.1 | 3859 | 68 | ||
| XM_021087443.1 | XP_020943102.1 | 3784 | 68 | |||
| XM_021087444.1 | XP_020943103.1 | 4492 | 68 | |||
| XM_021087445.1 | XP_020943104.1 | 3538 | 68 | |||
| GenBank mRNA | AK393132.1 | - | 1 | 1026 | 68 | |
More Information
Set Membership: |
|

SPRED2
Genomic Map