Assay Details
| Gene Symbol: HS3ST3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK393405.1 | - | 1 - 2 | 152 | 89 | |
| AK396553.1 | - | 1 - 2 | 132 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS3ST3A1| Gene Symbol: HS3ST3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK393405.1 | - | 1 - 2 | 152 | 89 | |
| AK396553.1 | - | 1 - 2 | 132 | 89 | ||
Set Membership: |
|