Assay Details
| Gene Symbol: SLC22A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122570.4 | XP_003122618.2 | 1418 | 61 | ||
| XM_005660741.3 | XP_005660798.2 | 1070 | 61 | |||
| XM_005660743.3 | XP_005660800.2 | 499 | 61 | |||
| XM_005660744.3 | XP_005660801.2 | 499 | 61 | |||
| XM_021082874.1 | XP_020938533.1 | 1398 | 61 | |||
| GenBank mRNA | AK393531.1 | - | 7 - 8 | 1022 | 61 | |
More Information
Set Membership: |
|

SLC22A12
Genomic Map