Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357423.4 | XP_003357471.1 | 2335 | 87 | ||
| XM_005667577.3 | XP_005667634.1 | 1623 | 87 | |||
| XM_005667578.3 | XP_005667635.1 | 1491 | 87 | |||
| XM_013979664.2 | XP_013835118.1 | 1417 | 87 | |||
| GenBank mRNA | AK393692.1 | - | 5 - 6 | 1467 | 87 | |
More Information
Set Membership: |
|

STEAP2
Genomic Map