Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067274.1 | XP_020922933.1 | 3834 | 88 | ||
| GenBank mRNA | AK394213.1 | - | 1 - 2 | 216 | 88 | |
More Information
Set Membership: |
|

RNF43
Genomic Map