Assay Details
| Gene Symbol: TIMMDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070323.1 | XP_020925982.1 | 578 | 65 | ||
| GenBank mRNA | AK394326.1 | - | 3 - 4 | 487 | 65 | |
More Information
Set Membership: |
|

TIMMDC1
Genomic Map