Assay Details
| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925334.3 | XP_001925369.1 | 1611 | 122 | ||
| GenBank mRNA | AK395339.1 | - | 1 - 2 | 623 | 122 | |
More Information
Set Membership: |
|

FBXO43
Genomic Map