Assay Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003484120.4 | XP_003484168.1 | 953 | 78 | ||
| XM_005673699.3 | XP_005673756.1 | 938 | 78 | |||
| GenBank mRNA | AK395713.1 | - | 6 - 7 | 859 | 78 | |
More Information
Set Membership: |
|

YIPF6
Genomic Map