Assay Details
| Gene Symbol: GATAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013979687.2 | XP_013835141.2 | 446 | 69 | ||
| XM_021102379.1 | XP_020958038.1 | 456 | 69 | |||
| GenBank mRNA | AK395771.1 | - | 1 - 2 | 294 | 69 | |
| AY609671.1 | - | 1 - 2 | 264 | 69 | ||
More Information
Set Membership: |
|

GATAD1
Genomic Map