Assay Details
| Gene Symbol: MED26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480761.4 | XP_003480809.1 | 435 | 66 | ||
| XM_005661191.3 | XP_005661248.1 | 421 | 66 | |||
| XM_021083584.1 | XP_020939243.1 | 4059 | 66 | |||
| GenBank mRNA | AK396350.1 | - | 2 - 3 | 396 | 66 | |
More Information
Set Membership: |
|

MED26
Genomic Map