Assay Details
| Gene Symbol: SESN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090855.1 | XP_020946514.1 | 1159 | 62 | ||
| XM_021090862.1 | XP_020946521.1 | 567 | 62 | |||
| XM_021090868.1 | XP_020946527.1 | 299 | 62 | |||
| XM_021090872.1 | XP_020946531.1 | 170 | 62 | |||
| GenBank mRNA | AK396502.1 | - | 2 - 3 | 241 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021090872.1 transcripts.
Set Membership: |
|

SESN1
Genomic Map