Assay Details
| Gene Symbol: ZNF185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986494.1 | XP_013841948.1 | 2111 | 73 | ||
| XM_013986496.2 | XP_013841950.1 | 2048 | 73 | |||
| XM_013986497.1 | XP_013841951.1 | 2027 | 73 | |||
| XM_013986498.1 | XP_013841952.1 | 2024 | 73 | |||
| XM_013986500.2 | XP_013841954.1 | 2003 | 73 | |||
| XM_013986501.2 | XP_013841955.1 | 2171 | 73 | |||
| XM_013986502.1 | XP_013841956.1 | 1937 | 73 | |||
| XM_013986504.1 | XP_013841958.1 | 1934 | 73 | |||
| XM_021079960.1 | XP_020935619.1 | 2113 | 73 | |||
| GenBank mRNA | AK397322.1 | - | 6 - 7 | 654 | 73 | |
More Information
Set Membership: |
|

ZNF185
Genomic Map