Assay Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129390.5 | XP_003129438.1 | 772 | 91 | ||
| XM_021062249.1 | XP_020917908.1 | 659 | 91 | |||
| XM_021062250.1 | XP_020917909.1 | 585 | 91 | |||
| GenBank mRNA | AK397536.1 | - | 4 - 5 | 389 | 91 | |
More Information
Set Membership: |
|

TMEM41B
Genomic Map