Assay Details
| Gene Symbol: FAM149A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984057.2 | XP_013839511.2 | 1597 | 69 | ||
| XM_013984058.2 | XP_013839512.2 | 992 | 69 | |||
| GenBank mRNA | AK398170.1 | - | 4 - 5 | 484 | 69 | |
More Information
Set Membership: |
|

FAM149A
Genomic Map