Assay Details
| Gene Symbol: FAM20A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656933.3 | XP_005656990.1 | 1688 | 70 | ||
| XM_021066714.1 | XP_020922373.1 | 1593 | 70 | |||
| XM_021066715.1 | XP_020922374.1 | 1691 | 70 | |||
| XM_021066716.1 | XP_020922375.1 | 600 | 70 | |||
| GenBank mRNA | AK398234.1 | - | 2 - 3 | 140 | 70 | |
More Information
Set Membership: |
|

FAM20A
Genomic Map