Assay Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088774.1 | XP_020944433.1 | 165 | 96 | ||
| GenBank mRNA | AK399767.1 | - | 1 - 2 | 81 | 96 | |
| AK400902.1 | - | 1 - 2 | 76 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021088774.1 transcripts.
Set Membership: |
|

LY6D
Genomic Map