Assay Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088774.1 | XP_020944433.1 | 265 | 81 | ||
| GenBank mRNA | AK399767.1 | - | 2 - 3 | 181 | 81 | |
| AK400902.1 | - | 2 - 3 | 176 | 81 | ||
More Information
Set Membership: |
|

LY6D
Genomic Map