Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088010.1 | XP_020943669.1 | 716 | 62 | ||
| GenBank mRNA | AY610207.1 | - | 4 - 5 | 628 | 62 | |
More Information
Set Membership: |
|

TMEM219
Genomic Map