Assay Details
Gene Symbol: CALHM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013992782.2 | XP_013848236.1 | 1899 | 78 | ||
XM_021091581.1 | XP_020947240.1 | 539 | 78 | |||
GenBank mRNA | AY610480.1 | - | 1 - 2 | 66 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
