Assay Details
| Gene Symbol: SPINK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661791.2 | XP_005661848.1 | 326 | 93 | ||
| GenBank mRNA | FM995240.1 | - | 2 - 3 | 167 | 93 | |
More Information
Set Membership: |
|

SPINK14
Genomic Map