Assay Details
| Gene Symbol: OSGIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925927.5 | XP_001925962.1 | 886 | 91 | ||
| XM_005662982.3 | XP_005663039.1 | 728 | 91 | |||
| XM_013996624.2 | XP_013852078.1 | 695 | 91 | |||
More Information
Set Membership: |
|

OSGIN2
Genomic Map