Assay Details
| Gene Symbol: BPIFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483940.4 | XP_003483988.2 | 1570 | 64 | ||
| XM_021078082.1 | XP_020933741.1 | 2266 | 64 | |||
More Information
Set Membership: |
|

BPIFB6
Genomic Map