Assay Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360737.4 | XP_003360785.2 | 724 | 64 | ||
| XM_021091896.1 | XP_020947555.1 | 412 | 64 | |||
| GenBank mRNA | AK232274.1 | - | 3 - 4 | 462 | 64 | |
| AK351220.1 | - | 3 - 4 | 414 | 64 | ||
| AK389418.1 | - | 3 - 4 | 482 | 64 | ||
More Information
Set Membership: |
|

TSPAN31
Genomic Map