Assay Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021085036.1 | XP_020940695.1 | 932 | 94 | ||
| XM_021085037.1 | XP_020940696.1 | 1016 | 94 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021085036.1 | XP_020940695.1 | 932 | 94 | ||
| XM_021085037.1 | XP_020940696.1 | 1016 | 94 | |||
Set Membership: |
|