Assay Details
| Gene Symbol: TMEM104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131226.4 | XP_003131274.1 | 395 | 60 | ||
| XM_005668622.3 | XP_005668679.1 | 360 | 60 | |||
| XM_005668623.3 | XP_005668680.1 | 360 | 60 | |||
More Information
Set Membership: |
|

TMEM104
Genomic Map