Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995291.2 | XP_013850745.1 | 584 | 107 | ||
| XM_013995305.2 | XP_013850759.1 | 547 | 107 | |||
| XM_013995314.2 | XP_013850768.1 | 581 | 107 | |||
| XM_021094920.1 | XP_020950579.1 | 2040 | 107 | |||
| XM_021094922.1 | XP_020950581.1 | 545 | 107 | |||
More Information
Set Membership: |
|

ZNF432
Genomic Map