Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013980996.2 | XP_013836450.2 | 1905 | 67 | ||
| XM_021067133.1 | XP_020922792.1 | 4919 | 67 | |||
| XM_021067134.1 | XP_020922793.1 | 1325 | 67 | |||
More Information
Set Membership: |
|

ZNF652
Genomic Map