Assay Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929174.7 | XP_001929209.4 | 493 | 99 | ||
| XM_021072447.1 | XP_020928106.1 | 249 | 99 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001929174.7, XM_021072447.1 transcripts.
Set Membership: |
|

FAM222A
Genomic Map