Assay Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929174.7 | XP_001929209.4 | 618 | 88 | ||
| XM_021072447.1 | XP_020928106.1 | 374 | 88 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM222A| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929174.7 | XP_001929209.4 | 618 | 88 | ||
| XM_021072447.1 | XP_020928106.1 | 374 | 88 | |||
Set Membership: |
|