Assay Details
| Gene Symbol: ZNF597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995710.2 | XP_013851164.1 | 1942 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013995710.2 transcripts.
Set Membership: |
|

ZNF597
Genomic Map