Assay Details
| Gene Symbol: ARRDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081097.1 | XP_020936756.1 | 794 | 69 | ||
| XM_021081098.1 | XP_020936757.1 | 793 | 69 | |||
| XM_021081099.1 | XP_020936758.1 | 694 | 69 | |||
| XM_021081100.1 | XP_020936759.1 | 717 | 69 | |||
| XM_021081101.1 | XP_020936760.1 | 717 | 69 | |||
| XM_021081102.1 | XP_020936761.1 | 1589 | 69 | |||
| XM_021081103.1 | XP_020936762.1 | 1408 | 69 | |||
| XM_021081104.1 | XP_020936763.1 | 1150 | 69 | |||
| XM_021081105.1 | XP_020936764.1 | 1007 | 69 | |||
| XM_021081106.1 | XP_020936765.1 | 711 | 69 | |||
| XM_021081107.1 | XP_020936766.1 | 622 | 69 | |||
| XM_021081108.1 | XP_020936767.1 | 890 | 69 | |||
| XM_021081109.1 | XP_020936768.1 | 1027 | 69 | |||
| XM_021081110.1 | XP_020936769.1 | 980 | 69 | |||
| XM_021081111.1 | XP_020936770.1 | 1025 | 69 | |||
| XM_021081112.1 | XP_020936771.1 | 825 | 69 | |||
| XM_021081113.1 | XP_020936772.1 | 571 | 69 | |||
| GenBank mRNA | KF589202.1 | - | 622 | 69 | ||
More Information
Set Membership: |
|

ARRDC1
Genomic Map