Assay Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355993.4 | XP_003356041.2 | 173 | 65 | ||
| XM_013998564.2 | XP_013854018.2 | 157 | 65 | |||
| XM_013998565.2 | XP_013854019.2 | 189 | 65 | |||
More Information
Set Membership: |
|

TMEM143
Genomic Map