Assay Details
| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005667897.2 | XP_005667954.1 | 156 | 99 | ||
| XM_013980004.2 | XP_013835458.1 | 486 | 99 | |||
| XM_013980005.2 | XP_013835459.1 | 211 | 99 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005667897.2, XM_013980004.2, XM_013980005.2 transcripts.
Set Membership: |
|

FIGNL1
Genomic Map