Assay Details
| Gene Symbol: ZGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101661.1 | XP_020957320.1 | 752 | 69 | ||
| XM_021101662.1 | XP_020957321.1 | 838 | 69 | |||
| XM_021101663.1 | XP_020957322.1 | 481 | 69 | |||
| XM_021101664.1 | XP_020957323.1 | 752 | 69 | |||
| XM_021101665.1 | XP_020957324.1 | 752 | 69 | |||
| XM_021101666.1 | XP_020957325.1 | 752 | 69 | |||
| XM_021101667.1 | XP_020957326.1 | 752 | 69 | |||
| XM_021101668.1 | XP_020957327.1 | 752 | 69 | |||
More Information
Set Membership: |
|

ZGRF1
Genomic Map