Assay Details
| Gene Symbol: DCLRE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926859.4 | XP_001926894.1 | 3484 | 78 | ||
| XM_013983742.2 | XP_013839196.1 | 3242 | 78 | |||
| XM_013983743.2 | XP_013839197.1 | 3253 | 78 | |||
| XM_013983744.2 | XP_013839198.1 | 3250 | 78 | |||
| XM_021073330.1 | XP_020928989.1 | 3221 | 78 | |||
More Information
Set Membership: |
|

DCLRE1A
Genomic Map