Assay Details
| Gene Symbol: HSDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126834.4 | XP_003126882.1 | 1058 | 68 | ||
| XM_005664318.3 | XP_005664375.1 | 1006 | 68 | |||
| XM_005664320.3 | XP_005664377.1 | 1017 | 68 | |||
| XM_013998155.2 | XP_013853609.1 | 960 | 68 | |||
More Information
Set Membership: |
|

HSDL1
Genomic Map