Assay Details
| Gene Symbol: HIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005669112.3 | XP_005669169.2 | 158 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005669112.3 transcripts.
Set Membership: |
|

HIC1
Genomic Map